Assay Details
| Gene Symbol: TMEM50A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261245.1 | NP_001248174.1 | 567 | 68 | ||
| XM_015131084.2 | XP_014986570.1 | 608 | 68 | |||
| GenBank mRNA | JU333738.1 | - | 584 | 68 | ||
| JU473274.1 | - | 588 | 68 | |||
| JV045508.1 | - | 567 | 68 | |||
More Information
Set Membership: |
|

TMEM50A
Genomic Map