Assay Details
| Gene Symbol: PIGC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261250.1 | NP_001248179.1 | 921 | 84 | ||
| XM_015123268.2 | XP_014978754.1 | 1653 | 84 | |||
| XM_028842585.1 | XP_028698418.1 | 3141 | 84 | |||
| XM_028842629.1 | XP_028698462.1 | 3137 | 84 | |||
| XM_028842676.1 | XP_028698509.1 | 3060 | 84 | |||
| XM_028842705.1 | XP_028698538.1 | 3100 | 84 | |||
| XM_028842735.1 | XP_028698568.1 | 2945 | 84 | |||
| GenBank mRNA | JU470844.1 | - | 1021 | 84 | ||
| JV043443.1 | - | 921 | 84 | |||
| JV047379.1 | - | 969 | 84 | |||
| JV047380.1 | - | 921 | 84 | |||
| JV047381.1 | - | 918 | 84 | |||
| JV047382.1 | - | 921 | 84 | |||
| JV635546.1 | - | 978 | 84 | |||
More Information
Set Membership: |
|

PIGC
Genomic Map