Assay Details
| Gene Symbol: RNF139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261771.1 | NP_001248700.1 | 499 | 106 | ||
| GenBank mRNA | JU321116.1 | - | 179 | 106 | ||
| JU473671.1 | - | 510 | 106 | |||
| JV043360.1 | - | 499 | 106 | |||
| JV639013.1 | - | 516 | 106 | |||
More Information
Set Membership: |
|

RNF139
Genomic Map