Assay Details
| Gene Symbol: RBM15B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265669.1 | NP_001252598.1 | 2725 | 65 | ||
| GenBank mRNA | JU326647.1 | - | 2018 | 65 | ||
| JV600275.1 | - | 2725 | 65 | |||
| JV713209.1 | - | 1808 | 65 | |||
More Information
Set Membership: |
|

RBM15B
Genomic Map