Assay Details
| Gene Symbol: RPP25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265677.1 | NP_001252606.1 | 419 | 87 | ||
| GenBank mRNA | JV585630.1 | - | 284 | 87 | ||
| JV600294.1 | - | 419 | 87 | |||
More Information
Set Membership: |
|

RPP25
Genomic Map