Assay Details
| Gene Symbol: ZSWIM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265702.1 | NP_001252631.1 | 378 | 63 | ||
| XM_015118658.1 | XP_014974144.1 | 418 | 63 | |||
| XM_015118660.2 | XP_014974146.1 | 318 | 63 | |||
| GenBank mRNA | JV635809.1 | - | 378 | 63 | ||
| JV706743.1 | - | 413 | 63 | |||
More Information
Set Membership: |
|

ZSWIM7
Genomic Map