Assay Details
| Gene Symbol: PTPMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265803.1 | NP_001252732.1 | 534 | 66 | ||
| GenBank mRNA | JV667338.1 | - | 534 | 66 | ||
| JV706336.1 | - | 526 | 66 | |||
More Information
Set Membership: |
|

PTPMT1
Genomic Map