Assay Details
| Gene Symbol: CLDN22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265844.1 | NP_001252773.1 | 623 | 73 | ||
| GenBank mRNA | JU335044.1 | - | 623 | 73 | ||
| Gene Symbol: WWC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015139578.2 | XP_014995064.2 | 8574 | 73 | ||
| XM_028848906.1 | XP_028704739.1 | 8793 | 73 | |||
| XM_028848907.1 | XP_028704740.1 | 8778 | 73 | |||
| XM_028848908.1 | XP_028704741.1 | 8709 | 73 | |||
| XM_028848909.1 | XP_028704742.1 | 8673 | 73 | |||
| XM_028848910.1 | XP_028704743.1 | 8589 | 73 | |||
More Information
Set Membership: |
|

CLDN22
Genomic Map