Assay Details
| Gene Symbol: CIBAR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265930.1 | NP_001252859.1 | 110 | 61 | ||
| XM_015145644.2 | XP_015001130.1 | 61 | ||||
| XM_015145648.2 | XP_015001134.1 | 61 | ||||
| XM_028852142.1 | XP_028707975.1 | 61 | ||||
| GenBank mRNA | JU321212.1 | - | 86 | 61 | ||
| JU473114.1 | - | 140 | 61 | |||
| JV044672.1 | - | 110 | 61 | |||
More Information
Set Membership: |
|

CIBAR1
Genomic Map