Assay Details
| Gene Symbol: FEN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265950.1 | NP_001252879.1 | 136 | 80 | ||
| GenBank mRNA | JU321864.1 | - | 115 | 80 | ||
| JU472990.1 | - | 115 | 80 | |||
| JV043309.1 | - | 136 | 80 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001265950.1 transcripts.
Set Membership: |
|

FEN1
Genomic Map