Assay Details
| Gene Symbol: VAMP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265988.1 | NP_001252917.1 | 207 | 78 | ||
| GenBank mRNA | JU323505.1 | - | 207 | 78 | ||
| JV705554.1 | - | 120 | 78 | |||
More Information
Set Membership: |
|

VAMP5
Genomic Map