Assay Details
| Gene Symbol: IER2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266014.1 | NP_001252943.1 | 314 | 88 | ||
| GenBank mRNA | JU477945.1 | - | 314 | 88 | ||
| JV634220.1 | - | 420 | 88 | |||
More Information
Set Membership: |
|

IER2
Genomic Map