Assay Details
| Gene Symbol: GPR37L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266071.1 | NP_001253000.1 | 699 | 78 | ||
| GenBank mRNA | JU321346.1 | - | 699 | 78 | ||
| JV598512.1 | - | 643 | 78 | |||
More Information
Set Membership: |
|

GPR37L1
Genomic Map