Assay Details
| Gene Symbol: GNG13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266642.1 | NP_001253571.1 | 107 | 106 | ||
| XM_028840598.1 | XP_028696431.1 | 277 | 106 | |||
| XM_028840600.1 | XP_028696433.1 | 255 | 106 | |||
| GenBank mRNA | JV667415.1 | - | 107 | 106 | ||
More Information
Set Membership: |
|

GNG13
Genomic Map