Assay Details
| Gene Symbol: TSPAN18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266877.1 | NP_001253806.1 | 455 | 62 | ||
| XM_015114242.2 | XP_014969728.1 | 739 | 62 | |||
| XM_028832768.1 | XP_028688601.1 | 946 | 62 | |||
| XM_028832769.1 | XP_028688602.1 | 808 | 62 | |||
| XM_028832770.1 | XP_028688603.1 | 1254 | 62 | |||
| XM_028832771.1 | XP_028688604.1 | 362 | 62 | |||
| XM_028832772.1 | XP_028688605.1 | 939 | 62 | |||
| XM_028832773.1 | XP_028688606.1 | 861 | 62 | |||
| XM_028832774.1 | XP_028688607.1 | 782 | 62 | |||
| XM_028832775.1 | XP_028688608.1 | 757 | 62 | |||
| GenBank mRNA | JU495829.1 | - | 145 | 62 | ||
| JV635503.1 | - | 455 | 62 | |||
More Information
Set Membership: |
|

TSPAN18
Genomic Map