Assay Details
| Gene Symbol: ZHX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267014.1 | NP_001253943.1 | 3042 | 111 | ||
| XM_015145938.2 | XP_015001424.1 | 3277 | 111 | |||
| GenBank mRNA | JU321442.1 | - | 3057 | 111 | ||
| JU321443.1 | - | 3057 | 111 | |||
| JU321445.1 | - | 3068 | 111 | |||
| JU472618.1 | - | 3094 | 111 | |||
| JV043528.1 | - | 3042 | 111 | |||
More Information
Set Membership: |
|

ZHX1
Genomic Map