Assay Details
| Gene Symbol: COA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267542.1 | NP_001254471.1 | 281 | 116 | ||
| GenBank mRNA | JU334707.1 | - | 278 | 116 | ||
| JV045604.1 | - | 281 | 116 | |||
More Information
Set Membership: |
|

COA5
Genomic Map