Assay Details
| Gene Symbol: GNG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267786.1 | NP_001254715.1 | 282 | 74 | ||
| GenBank mRNA | JU319692.1 | - | 282 | 74 | ||
| JV637485.1 | - | 106 | 74 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001267786.1 transcripts.
Set Membership: |
|

GNG3
Genomic Map