Assay Details
| Gene Symbol: HIST3H2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265626.1 | NP_001252555.1 | 121 | 89 | ||
| GenBank mRNA | JV564973.1 | - | 121 | 89 | ||
| Gene Symbol: H2AFX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JV713332.1 | - | 118 | 89 | ||
| Gene Symbol: HIST3H2BB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028826626.1 | XP_028682459.1 | 755 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028826626.1 transcripts.
Set Membership: |
|

HIST3H2A
Genomic Map