Assay Details
| Gene Symbol: MRPS17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266196.2 | NP_001253125.1 | 134 | 109 | ||
| XM_015133415.2 | XP_014988901.1 | 248 | 109 | |||
| XM_015133416.2 | XP_014988902.1 | 445 | 109 | |||
| XM_015133417.2 | XP_014988903.1 | 213 | 109 | |||
| GenBank mRNA | JU321888.1 | - | 137 | 109 | ||
| JU472001.1 | - | 142 | 109 | |||
| JV043524.1 | - | 137 | 109 | |||
More Information
Set Membership: |
|

MRPS17
Genomic Map