Assay Details
| Gene Symbol: REG1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266520.2 | NP_001253449.1 | 375 | 129 | ||
| XM_015112723.2 | XP_014968209.1 | 368 | 129 | |||
| GenBank mRNA | JU335778.1 | - | 375 | 129 | ||
More Information
Set Membership: |
|

REG1A
Genomic Map