Assay Details
| Gene Symbol: CRABP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266889.1 | NP_001253818.1 | 373 | 110 | ||
| GenBank mRNA | JV457355.1 | - | 477 | 110 | ||
| JV667312.1 | - | 373 | 110 | |||
More Information
Set Membership: |
|

CRABP2
Genomic Map