Assay Details
| Gene Symbol: TRIM68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260732.1 | NP_001247661.1 | 908 | 129 | ||
| XM_028832813.1 | XP_028688646.1 | 977 | 129 | |||
| GenBank mRNA | JU335234.1 | - | 979 | 129 | ||
| JU476200.1 | - | 908 | 129 | |||
| JV064715.1 | - | 1009 | 129 | |||
More Information
Set Membership: |
|

TRIM68
Genomic Map