Assay Details
| Gene Symbol: SRP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265919.1 | NP_001252848.1 | 1013 | 103 | ||
| GenBank mRNA | JV048029.1 | - | 1013 | 103 | ||
| JV721882.1 | - | 620 | 103 | |||
More Information
Set Membership: |
|

SRP14
Genomic Map