Assay Details
| Gene Symbol: SUMO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261108.1 | NP_001248037.1 | 817 | 84 | ||
| GenBank mRNA | JU476899.1 | - | 756 | 84 | ||
| JV044144.1 | - | 817 | 84 | |||
| JV667291.1 | - | 818 | 84 | |||
More Information
Set Membership: |
|

SUMO3
Genomic Map