Assay Details
| Gene Symbol: CAPZA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261214.1 | NP_001248143.1 | 1521 | 174 | ||
| XM_015147087.2 | XP_015002573.1 | 1123 | 177 | |||
| GenBank mRNA | JU336587.1 | - | 1322 | 173 | ||
| JV045517.1 | - | 1521 | 174 | |||
More Information
Set Membership: |
|

CAPZA1
Genomic Map