Assay Details
| Gene Symbol: ST13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261322.1 | NP_001248251.1 | 2022 | 75 | ||
| GenBank mRNA | JU336079.1 | - | 2087 | 75 | ||
| JU471405.1 | - | 1993 | 75 | |||
| JV043930.1 | - | 2022 | 75 | |||
More Information
Set Membership: |
|

ST13
Genomic Map