Assay Details
| Gene Symbol: SERPINB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202551.2 | NP_001189480.1 | 788 | 106 | ||
| XM_015135413.2 | XP_014990899.2 | 106 | ||||
| XM_015135414.2 | XP_014990900.1 | 106 | ||||
| XM_015135415.2 | XP_014990901.1 | 106 | ||||
| XM_015135416.2 | XP_014990902.1 | 106 | ||||
| XM_028846839.1 | XP_028702672.1 | 106 | ||||
| GenBank mRNA | EG702332.1 | - | 106 | |||
| JU334995.1 | - | 106 | ||||
| JV044290.1 | - | 106 | ||||
More Information
Set Membership: |
|

SERPINB6
Genomic Map