Assay Details
| Gene Symbol: MESD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015142865.2 | XP_014998351.1 | 327 | 99 | ||
| GenBank mRNA | JU322959.1 | - | 240 | 99 | ||
| JU472547.1 | - | 232 | 99 | |||
| JV043663.1 | - | 243 | 99 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MESD| Gene Symbol: MESD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015142865.2 | XP_014998351.1 | 327 | 99 | ||
| GenBank mRNA | JU322959.1 | - | 240 | 99 | ||
| JU472547.1 | - | 232 | 99 | |||
| JV043663.1 | - | 243 | 99 | |||