Assay Details
| Gene Symbol: DNASE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015123039.2 | XP_014978525.2 | 995 | 58 | ||
| GenBank mRNA | JU323447.1 | - | 799 | 58 | ||
| JU481702.1 | - | 603 | 58 | |||
More Information
Set Membership: |
|

DNASE2
Genomic Map