Assay Details
| Gene Symbol: RPL22L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015131627.2 | XP_014987113.1 | 530 | 115 | ||
| XM_015131628.2 | XP_014987114.1 | 291 | 115 | |||
| GenBank mRNA | JU471798.1 | - | 302 | 115 | ||
| JV054180.1 | - | 210 | 115 | |||
More Information
Set Membership: |
|

RPL22L1
Genomic Map