Assay Details
| Gene Symbol: MTFMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015142472.2 | XP_014997958.2 | 990 | 96 | ||
| GenBank mRNA | JU338630.1 | - | 855 | 96 | ||
| JV051414.1 | - | 190 | 96 | |||
More Information
Set Membership: |
|

MTFMT
Genomic Map