Assay Details
| Gene Symbol: YRDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002802325.3 | XP_002802371.2 | 810 | 74 | ||
| XM_015135752.2 | XP_014991238.1 | 665 | 74 | |||
| GenBank mRNA | JU327195.1 | - | 602 | 74 | ||
| JU327196.1 | - | 602 | 74 | |||
| JV661521.1 | - | 644 | 74 | |||
More Information
Set Membership: |
|

YRDC
Genomic Map