Assay Details
Gene Symbol: PPP4R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002802736.3 | XP_002802782.1 | 741 | 76 | ||
XM_028844366.1 | XP_028700199.1 | 570 | 76 | |||
GenBank mRNA | JU322478.1 | - | 554 | 76 | ||
JU472278.1 | - | 560 | 76 | |||
JV045383.1 | - | 560 | 76 | |||
JV668825.1 | - | 563 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
