Assay Details
Gene Symbol: UNC93A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015137681.2 | XP_014993167.2 | 2141 | 67 | ||
XM_028846974.1 | XP_028702807.1 | 492 | 67 | |||
XM_028846975.1 | XP_028702808.1 | 465 | 67 | |||
XM_028846977.1 | XP_028702810.1 | 2144 | 67 | |||
XM_028846978.1 | XP_028702811.1 | 2144 | 67 | |||
XM_028846979.1 | XP_028702812.1 | 2143 | 67 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028846978.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
