Assay Details
| Gene Symbol: MRPL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015122841.2 | XP_014978327.1 | 350 | 50 | ||
| XM_015122842.2 | XP_014978328.1 | 358 | 50 | |||
| GenBank mRNA | JU487632.1 | - | 322 | 50 | ||
| JU487635.1 | - | 322 | 50 | |||
More Information
Set Membership: |
|

MRPL4
Genomic Map