Assay Details
Gene Symbol: CEACAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015124068.2 | XP_014979554.2 | 1560 | 62 | ||
XM_028838885.1 | XP_028694718.1 | 1548 | 62 | |||
XM_028838887.1 | XP_028694720.1 | 1272 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
