Assay Details
| Gene Symbol: SRP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015115259.1 | XP_014970745.1 | 207 | 84 | ||
| GenBank mRNA | JU336065.1 | - | 199 | 84 | ||
| JU347637.1 | - | 199 | 84 | |||
| JU476933.1 | - | 195 | 84 | |||
More Information
Set Membership: |
|

SRP9
Genomic Map