Assay Details
| Gene Symbol: PIGZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015132598.2 | XP_014988084.2 | 306 | 68 | ||
| XM_028843545.1 | XP_028699378.1 | 400 | 68 | |||
| XM_028843546.1 | XP_028699379.1 | 401 | 68 | |||
| XM_028843547.1 | XP_028699380.1 | 356 | 68 | |||
| GenBank mRNA | JU325068.1 | - | 155 | 68 | ||
| JV521308.1 | - | 206 | 68 | |||
| JV594546.1 | - | 145 | 68 | |||
More Information
Set Membership: |
|

PIGZ
Genomic Map