Assay Details
| Gene Symbol: ANKRD61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133263.2 | XP_014988749.2 | 378 | 92 | ||
| XM_028845140.1 | XP_028700973.1 | 376 | 92 | |||
| XM_028845141.1 | XP_028700974.1 | 299 | 92 | |||
| XM_028845142.1 | XP_028700975.1 | 296 | 92 | |||
More Information
Set Membership: |
|

ANKRD61
Genomic Map