Assay Details
| Gene Symbol: THNSL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015112817.2 | XP_014968303.2 | 533 | 61 | ||
| XM_015112820.2 | XP_014968306.2 | 2769 | 61 | |||
| XM_028832316.1 | XP_028688149.1 | 817 | 61 | |||
| XM_028832317.1 | XP_028688150.1 | 789 | 61 | |||
| XM_028832318.1 | XP_028688151.1 | 531 | 61 | |||
| GenBank mRNA | JU325743.1 | - | 410 | 61 | ||
| JV044287.1 | - | 434 | 61 | |||
| JV473425.1 | - | 448 | 61 | |||
More Information
Set Membership: |
|

THNSL2
Genomic Map