Assay Details
| Gene Symbol: ESRRA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015113888.2 | XP_014969374.1 | 1153 | 65 | ||
| XM_015113889.2 | XP_014969375.1 | 984 | 65 | |||
| XM_015113890.2 | XP_014969376.2 | 834 | 65 | |||
| XM_028833298.1 | XP_028689131.1 | 984 | 65 | |||
| GenBank mRNA | JU333359.1 | - | 734 | 65 | ||
| JU474656.1 | - | 740 | 65 | |||
| JV574268.1 | - | 734 | 65 | |||
More Information
Set Membership: |
|

ESRRA
Genomic Map