Assay Details
| Gene Symbol: MXD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015141504.2 | XP_014996990.2 | 1789 | 75 | ||
| XM_028849945.1 | XP_028705778.1 | 1726 | 75 | |||
| XM_028849946.1 | XP_028705779.1 | 948 | 75 | |||
| XM_028849947.1 | XP_028705780.1 | 1783 | 75 | |||
| GenBank mRNA | JU471564.1 | - | 534 | 75 | ||
More Information
Set Membership: |
|

MXD3
Genomic Map