Assay Details
| Gene Symbol: JAKMIP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015148474.2 | XP_015003960.2 | 1876 | 57 | ||
| XM_028827023.1 | XP_028682856.1 | 1885 | 57 | |||
| XM_028827024.1 | XP_028682857.1 | 1889 | 57 | |||
| XM_028827025.1 | XP_028682858.1 | 1883 | 57 | |||
| XM_028827026.1 | XP_028682859.1 | 1883 | 57 | |||
| XM_028827027.1 | XP_028682860.1 | 1876 | 57 | |||
| XM_028827028.1 | XP_028682861.1 | 1660 | 57 | |||
| XM_028827029.1 | XP_028682862.1 | 1653 | 57 | |||
| XM_028827030.1 | XP_028682863.1 | 1657 | 57 | |||
| GenBank mRNA | JU331071.1 | - | 1464 | 57 | ||
| JU342026.1 | - | 1464 | 57 | |||
More Information
Set Membership: |
|

JAKMIP3
Genomic Map