Assay Details
| Gene Symbol: DHRS7C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015118594.2 | XP_014974080.2 | 868 | 73 | ||
| XM_015118595.2 | XP_014974081.2 | 865 | 73 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DHRS7C| Gene Symbol: DHRS7C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015118594.2 | XP_014974080.2 | 868 | 73 | ||
| XM_015118595.2 | XP_014974081.2 | 865 | 73 | |||
Set Membership: |
|