Assay Details
| Gene Symbol: ADH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001105718.4 | XP_001105718.2 | 1095 | 64 | ||
| GenBank mRNA | JV046187.1 | - | 1009 | 64 | ||
| JV046188.1 | - | 1009 | 64 | |||
| JV642224.1 | - | 1031 | 64 | |||
| JV642225.1 | - | 1031 | 64 | |||
More Information
Set Membership: |
|

ADH5
Genomic Map