Assay Details
| Gene Symbol: BCL7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028830020.1 | XP_028685853.1 | 1227 | 83 | ||
| XM_028830021.1 | XP_028685854.1 | 540 | 83 | |||
| GenBank mRNA | JU321668.1 | - | 258 | 83 | ||
| JU472189.1 | - | 272 | 83 | |||
| JU472190.1 | - | 272 | 83 | |||
| JV589890.1 | - | 327 | 83 | |||

BCL7A
Genomic Map