Assay Details
| Gene Symbol: ABCA15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125772.2 | XP_014981258.2 | 1564 | 62 | ||
| XM_028841598.1 | XP_028697431.1 | 1616 | 62 | |||
| XM_028841599.1 | XP_028697432.1 | 1447 | 62 | |||
| XM_028841600.1 | XP_028697433.1 | 1564 | 62 | |||
More Information
Set Membership: |
|

ABCA15
Genomic Map