Assay Details
| Gene Symbol: TAF6L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015113954.2 | XP_014969440.1 | 1608 | 60 | ||
| GenBank mRNA | JU321650.1 | - | 324 | 60 | ||
| JU473170.1 | - | 247 | 60 | |||
| JV045479.1 | - | 321 | 60 | |||
More Information
Set Membership: |
|

TAF6L
Genomic Map