Assay Details
| Gene Symbol: LOC719436 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015124675.2 | XP_014980161.2 | 245 | 73 | ||
| XM_015124677.2 | XP_014980163.2 | 245 | 73 | |||
More Information
Set Membership: |
|

LOC719436
Genomic Map